cis-Jasmone 95%

Supplier: Ambeed
Warning

Synonyms: (Z)-3-Methyl-2-(pent-2-en-1-yl)cyclopent-2-enone, cis-Jasmon

A219384-10G A219384-25G A219384-5G A219384-1G A219384-500G
103818-788EA 9 USD
103818-788 103849-664 76691-748 77088-218 77088-220 77303-526 77626-108
cis-Jasmone 95%
cis-Jasmone

Precaución: for Research Use Only

Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Boiling Pt: 233.2 °C (760 mm Hg)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00001402
CAS Number: 488-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR