(trans,trans)-4'-Propyl-[1,1'-bi(cyclohexane)]-4-carbonitrile 97%

Supplier: Ambeed
Warning

A549286-10G A549286-1G A549286-5G
103839-806EA 48 USD
103839-806 76774-814 76774-816 76843-884
(trans,trans)-4'-Propyl-[1,1'-bi(cyclohexane)]-4-carbonitrile 97%
(trans,trans)-4'-Propyl-[1,1'-bi(cyclohexane)]-4-carbonitrile

Precaución: for Research Use Only

Formula: C₁₆H₂₇N
MW: 233.39 g/mol
Boiling Pt: 353.3 °C (760 mm Hg)
Flash Pt: 155.2 °C
Storage Temperature: Ambient
MDL Number: MFCD11053435
CAS Number: 65355-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR