3-Aminomethyltetrahydrofuran 97%

Supplier: Ambeed
Danger

Synonyms: (Tetrahydrofuran-3-yl)methanamine, 3-(Aminomethyl)oxolane, (tetrahydrofuran-3-yl)methanamine, Oxolan-3-ylmethanamine, 3-(Aminomethyl)tetrahydrofuran

A401519-1G A401519-25G A401519-5G
103827-054EA 5 USD
103827-054 103838-090 76841-548 76841-550 76841-552
3-Aminomethyltetrahydrofuran 97%
3-Aminomethyltetrahydrofuran

Precaución: for Research Use Only

Formula: C₅H₁₁NO
Boiling Pt: 156 °C (760 mm Hg)
Flash Pt: 58.9 °C
Storage Temperature: Ambient
MDL Number: MFCD08234925
CAS Number: 165253-31-6
UN: 2924
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR