Potassium-4-(trifluoromethyl)phenyltrifluoroborate 97%

Supplier: Ambeed
Warning

Synonyms: Potassium 4-(trifluoromethyl)phenyltrifluoroborate, (T-4)-Potassium trifluoro(4-(trifluoromethyl)phenyl)borate

A107725-10G A107725-250MG A107725-25G A107725-1G A107725-5G
103797-240EA 6 USD
103797-240 103823-008 76717-912 76717-914 76717-916 77309-864 77309-866
Potassium-4-(trifluoromethyl)phenyltrifluoroborate 97%
Potassium 4-(trifluoromethyl)phenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₇H₄BF₆K
MW: 252.01 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD04039955
CAS Number: 166328-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR