(1S,2S)-(+)-1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)cobalt(II) 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S,S)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II)
A160042-10G
A160042-250MG
A160042-1G
A160042-25G
A160042-5G
A160042-100G
103805-322EA
6
USD
103805-322
103843-552
77191-050
77191-052
77319-462
77319-464
77319-466
77615-306
(1S,2S)-(+)-1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)cobalt(II) 99%
(1S,2S)-(+)-1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)cobalt(II)
Precaución: Sealed in dry, at room temperature.
Formula:
C₃₆H₅₂CoN₂O₂ Melting Pt: 409…412 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631278 CAS Number: 188264-84-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...