Chloro{[(1S,2S)-(+)-2-amino-1,2-diphenylethyl](4-toluenesulfonyl)amido}(p-cymene)ruthenium(II) 97%

Supplier: Ambeed
Warning

Synonyms: Chloro(p-cymene)[(S,S)-N-(p-toluenesulphonyl)-1,2-diphenylethylenediamine]ruthenium(II), (S,S)-N-(p-Toluenesulphonyl)-1,2-diphenylethanediamine(chloro)(p-cymene)ruthenium(II), [(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulphonamide]chloro(p-cymene)ruthenium(II), RuCl[(S,S)-Tsdpen](p-cymene)

A826324-250MG A826324-1G A826324-10G A826324-100G
103841-156EA 72 USD
103841-156 77036-564 77308-410 77573-638 77674-224
Chloro{[(1S,2S)-(+)-2-amino-1,2-diphenylethyl](4-toluenesulfonyl)amido}(p-cymene)ruthenium(II) 97%
Chloro{[(1S,2S)-(+)-2-amino-1,2-diphenylethyl](4-toluenesulfonyl)amido}(p-cymene)ruthenium(II)

Precaución: for Research Use Only

Formula: C₃₁H₃₅ClN₂O₂RuS
Melting Pt: >175 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09953484
CAS Number: 192139-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR