(1S,6S)-2,8-Diazabicyclo[4.3.0]nonane 97%

Supplier: Ambeed
Danger

A101196-100G A101196-25G A101196-1G A101196-5G
103795-852EA 11 USD
103795-852 103821-536 76994-492 76994-494 77297-018 77297-020
(1S,6S)-2,8-Diazabicyclo[4.3.0]nonane 97%
(1S,6S)-2,8-Diazabicyclo[4.3.0]nonane

Precaución: for Research Use Only

Formula: C₇H₁₄N₂
Boiling Pt: 285.8 °C (760 mm Hg)
Flash Pt: 126.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08458306
CAS Number: 151213-40-0
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR