(S)-Tetrahydrofuran-3-amine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A188717-1G
A188717-5G
A188717-100G
A188717-10G
A188717-25G
103816-124EA
8
USD
103816-124
103846-716
77084-470
77084-472
77191-900
77191-902
77191-904
77302-142
(S)-Tetrahydrofuran-3-amine hydrochloride 95%
(S)-Tetrahydrofuran-3-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₁₀ClNO Melting Pt: 160…163 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08445642 CAS Number: 204512-95-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...