(S)-Tetrahydro-2H-pyran-3-amine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A143846-100MG
A143846-250MG
A143846-1G
A143846-5G
103803-326EA
72
USD
103803-326
77027-732
77027-734
77316-632
77316-634
(S)-Tetrahydro-2H-pyran-3-amine hydrochloride 97%
(S)-Tetrahydro-2H-pyran-3-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₁NO·ClH MW: 137.608 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09056862 CAS Number: 1071829-81-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...