(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl pyrrolidin-3-ylcarbamate, (3S)-(-)-3-(Boc-amino)pyrrolidine

A107505-100G A107505-10G A107505-25G A107505-1G A107505-5G
103797-202EA 5 USD
103797-202 103822-970 76687-286 76687-288 76687-290 77309-828 77309-830
(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine 97%
(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₂
MW: 186.26 g/mol
Melting Pt: 52…56 °C
Flash Pt: 127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00143194
CAS Number: 122536-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR