(S)-tert-Butylmethyl(piperidin-3-yl)carbamate 97%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl methyl(piperidin-3-yl)carbamate

A226260-100MG A226260-250MG A226260-1G A226260-25G A226260-5G
103819-236EA 28 USD
103819-236 103850-136 77098-102 77098-104 77303-762 77303-764 77303-766
(S)-tert-Butylmethyl(piperidin-3-yl)carbamate 97%
(S)-tert-Butylmethyl(piperidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
Boiling Pt: 287.8 °C (760 mm Hg)
Flash Pt: 128 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09878597
CAS Number: 309962-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR