(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl 5-oxopyrrolidine-2-carboxylate, tert-Butyl L-Pyroglutamate, tert-Butyl (S)-5-oxopyrrolidine-2-carboxylate, tert-Butyl (S)-2-pyrrolidone-5-carboxylate

A161921-1G A161921-5G A161921-10G
103805-552EA 5 USD
103805-552 103843-796 77319-814 77319-816 77685-328
(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate 97%
(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
Boiling Pt: 319.2 °C (760 mm Hg)
Melting Pt: 91…92 °C
Storage Temperature: Ambient
MDL Number: MFCD06659481
CAS Number: 35418-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR