(S)-tert-Butyl 3-methyl-1,4-diazepane-1-carboxylate 95%

Supplier: Ambeed
Warning

A170499-5G A170499-10G A170499-25G
103806-508EA 18 USD
103806-508 103844-782 77081-894 77096-494 77096-496
(S)-tert-Butyl 3-methyl-1,4-diazepane-1-carboxylate 95%
(S)-tert-Butyl 3-methyl-1,4-diazepane-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.3 g/mol
Boiling Pt: 288.33 °C (760 mm Hg)
Flash Pt: 128.177 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09840678
CAS Number: 194032-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR