(S)-1-Boc-3-ethylpiperazine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl 3-ethylpiperazine-1-carboxylate

A261941-100MG A261941-250MG A261941-1G
103812-548EA 59 USD
103812-548 103852-440 77193-846 77193-848 77304-838
(S)-1-Boc-3-ethylpiperazine 97%
(S)-1-Boc-3-ethylpiperazine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.3046 g/mol
Boiling Pt: 287 °C (760 mm Hg)
Flash Pt: 127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07772110
CAS Number: 928025-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR