(S)-tert-Butyl-3-cyanopyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: (S)-1-Boc-3-cyanopyrrolidine, (S)-tert-Butyl 3-cyanopyrrolidine-1-carboxylate

A249995-100MG A249995-250MG A249995-1G
103811-906EA 38 USD
103811-906 103851-746 76692-462 76692-464 77304-512
(S)-tert-Butyl-3-cyanopyrrolidine-1-carboxylate 97%
(S)-tert-Butyl-3-cyanopyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₂
MW: 196.2500 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03419277
CAS Number: 132945-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR