(S)-(+)-3-Amino-1-boc-piperidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-1-Boc-3-aminopiperidine, (S)-tert-Butyl 3-aminopiperidine-1-carboxylate, (S)-(+)-3-Amino-1-BOC-piperidine
A525647-100G
A525647-10G
A525647-1G
A525647-25G
A525647-5G
103828-532EA
5
USD
103828-532
103839-646
76774-680
76774-682
76843-656
76843-658
76843-660
(S)-(+)-3-Amino-1-boc-piperidine 98%
(S)-(+)-3-Amino-1-Boc-piperidine
Precaución: for Research Use Only
Formula:
C₁₀H₂₀N₂O₂ Boiling Pt: 277.3 °C (760 mm Hg) Flash Pt: 121.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03094718 CAS Number: 625471-18-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...