(S)-tert-Butyl 3-(piperazin-1-yl)pyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A271341-100MG A271341-250MG
103813-044EA 480 USD
103813-044 103852-954 77014-422 77014-424
(S)-tert-Butyl 3-(piperazin-1-yl)pyrrolidine-1-carboxylate 97%
(S)-tert-Butyl 3-(piperazin-1-yl)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₅N₃O₂
MW: 255.36 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18633366
CAS Number: 1010446-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR