(S)-(+)-1-Boc-3-(methylamino)piperidine 95%

Supplier: Ambeed
Danger

Synonyms: (S)-tert-Butyl 3-(methylamino)piperidine-1-carboxylate

A349630-250MG A349630-1G A349630-25G
103826-022EA 40 USD
103826-022 103837-016 76648-462 76840-112 77748-686
(S)-(+)-1-Boc-3-(methylamino)piperidine 95%
(S)-(+)-1-Boc-3-(methylamino)piperidine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.309 g/mol
Boiling Pt: 340.4 °C (760 mm Hg)
Flash Pt: 159.694 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09752964
CAS Number: 912368-73-1
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR