(S)-1-Boc-3-(hydroxymethyl)piperazine 98%

Supplier: Ambeed
Warning

A109002-100G A109002-10G A109002-250MG A109002-25G A109002-1G A109002-5G
103797-422EA 10 USD
103797-422 103823-208 77093-854 77093-856 77093-858 77093-860 77310-042 77310-044
(S)-1-Boc-3-(hydroxymethyl)piperazine 98%
(S)-1-Boc-3-(hydroxymethyl)piperazine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₃
Boiling Pt: 322.9 °C (760 mm Hg)
Melting Pt: 110…114 °C
Flash Pt: 149.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05863889
CAS Number: 314741-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR