(S)-tert-Butyl 3-((methylsulfonyl)oxy)pyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A229347-250MG A229347-1G A229347-5G
103819-468EA 10 USD
103819-468 103850-368 77013-482 77089-204 77089-206
(S)-tert-Butyl 3-((methylsulfonyl)oxy)pyrrolidine-1-carboxylate 97%
(S)-tert-Butyl 3-((methylsulfonyl)oxy)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₅S
MW: 265.33 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09953436
CAS Number: 132945-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR