(S)-1-Boc-2-phenylpiperazine 95%

Supplier: Ambeed
Warning

A117495-100MG A117495-250MG A117495-5G
103798-900EA 135 USD
103798-900 77009-252 77009-254 77889-807
(S)-1-Boc-2-phenylpiperazine 95%
(S)-1-Boc-2-phenylpiperazine

Precaución: for Research Use Only

Formula: C₁₅H₂₂N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD08685943
CAS Number: 1240583-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR