(S)-1-Boc-2-methylpiperazine 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Methyl-1-piperazinecarboxylic acid tert-Butyl ester, tert-Butyl (S)-2-Methyl-1-piperazinecarboxylate, (S)-tert-Butyl 2-methylpiperazine-1-carboxylate, (S)-1-Boc-2-methylpiperazine, (S)-1-(tert-Butoxycarbonyl)-2-methylpiperazine

A154220-100G A154220-10G A154220-250MG A154220-500G A154220-25G A154220-1G A154220-5G
103804-614EA 6 USD
103804-614 103842-798 76689-474 76689-476 76689-478 76689-480 77300-044 77318-498 77318-500
(S)-1-Boc-2-methylpiperazine 98%
(S)-1-Boc-2-methylpiperazine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
Flash Pt: 116.33 °C
Storage Temperature: Ambient
MDL Number: MFCD01862121
CAS Number: 169447-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR