(S)-tert-Butyl 2-amino-3,3-dimethylbutanoate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-Tle-OtBu·HCl

A206633-1G A206633-5G A206633-250MG
103817-722EA 27 USD
103817-722 103848-398 77086-656 77086-658 77139-530
(S)-tert-Butyl 2-amino-3,3-dimethylbutanoate hydrochloride 97%
(S)-tert-Butyl 2-amino-3,3-dimethylbutanoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₂₂ClNO₂
MW: 223.74 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038904
CAS Number: 119483-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR