(S)-N-Boc-2-hydroxymethylmorpholine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-tert-Butyl 2-(hydroxymethyl)morpholine-4-carboxylate, (2S)-2-(Hydroxymethyl)morpholine, N-BOC protected
A327849-1G
A327849-25G
A327849-5G
A327849-100G
A327849-10G
A327849-250MG
103825-478EA
6
USD
103825-478
103836-428
76839-392
76839-394
76839-396
77194-938
77194-940
77194-942
(S)-N-Boc-2-hydroxymethylmorpholine 97%
(S)-N-Boc-2-hydroxymethylmorpholine
Precaución: for Research Use Only
Formula:
C₁₀H₁₉NO₄ MW: 217.265 g/mol Boiling Pt: 320.7 °C (760 mm Hg) Melting Pt: 62 °C Flash Pt: 148 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09260606 CAS Number: 135065-76-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...