(S)-4-Boc-2-(aminomethyl)morpholine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl 2-(aminomethyl)morpholine-4-carboxylate

A163236-100MG A163236-250MG A163236-1G A163236-5G A163236-25G
103805-702EA 65 USD
103805-702 103843-960 76783-958 76783-960 77320-052 77320-054 77871-821
(S)-4-Boc-2-(aminomethyl)morpholine 97%
(S)-4-Boc-2-(aminomethyl)morpholine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₃
MW: 216.277 g/mol
Boiling Pt: 310.7 °C (760 mm Hg)
Flash Pt: 141 °C
Storage Temperature: Ambient
MDL Number: MFCD11707017
CAS Number: 879403-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR