(S)-tert-Butyl 2-(2-hydroxyethyl)piperazine-1-carboxylate 95%

Supplier: Ambeed
Warning

A249455-100MG A249455-250MG
103811-870EA 618 USD
103811-870 103851-708 77193-626 77193-628
(S)-tert-Butyl 2-(2-hydroxyethyl)piperazine-1-carboxylate 95%
(S)-tert-Butyl 2-(2-hydroxyethyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₃
MW: 230.3 g/mol
Boiling Pt: 339.1 °C (760 mm Hg)
Flash Pt: 158.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10566720
CAS Number: 169448-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR