(S)-tert-Butyl (piperidin-2-ylmethyl)carbamate 95%

Supplier: Ambeed
Warning

A125234-100MG A125234-250MG A125234-1G
103800-648EA 96 USD
103800-648 76810-834 76810-836 77312-796
(S)-tert-Butyl (piperidin-2-ylmethyl)carbamate 95%
(S)-tert-Butyl (piperidin-2-ylmethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11975335
CAS Number: 139004-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR