(S)-tert-Butyl (2-oxopyrrolidin-3-yl)carbamate 97%

Supplier: Ambeed
Warning

A136483-100MG A136483-250MG A136483-1G A136483-5G
103802-306EA 139 USD
103802-306 77190-200 77190-202 77315-200 77315-202
(S)-tert-Butyl (2-oxopyrrolidin-3-yl)carbamate 97%
(S)-tert-Butyl (2-oxopyrrolidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₆N₂O₃
MW: 200.24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03426144
CAS Number: 92235-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR