N-(tert-Butoxycarbonyl)-L-2-phenylglycinol 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Phenylglycinol, (S)-tert-Butyl (2-hydroxy-1-phenylethyl)carbamate, N-Boc-L-2-phenylglycinol, (S)-2-(tert-Butoxycarbonylamino)-2-phenylethanol

A117390-100G A117390-10G A117390-25G A117390-500G A117390-1G A117390-5G
103798-876EA 5 USD
103798-876 103824-760 77009-242 77009-244 77009-246 77009-248 77311-536 77311-538
N-(tert-Butoxycarbonyl)-L-2-phenylglycinol 97%
N-(tert-Butoxycarbonyl)-L-2-phenylglycinol

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₃
Melting Pt: 136…139 °C
Flash Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00274206
CAS Number: 117049-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR