Boc-L-alaninal 97%

Supplier: Ambeed
Warning

Synonyms: N-tert-Butoxycarbonyl-L-alaninal, Boc-L-Ala-aldehyde, Boc-L-alaninal, Boc-Ala-aldehyde, (S)-(1-Oxopropan-2-yl)carbamic acid tert-Butyl ester, (S)-tert-Butyl (1-Oxopropan-2-yl)carbamate

A141806-100MG A141806-1G A141806-5G A141806-250MG A141806-100G
103803-064EA 88 USD
103803-064 76689-036 77299-246 77299-248 77600-058 77876-975
Boc-L-alaninal 97%
Boc-L-alaninal

Precaución: for Research Use Only

Formula: C₈H₁₅NO₃
MW: 173.212 g/mol
Boiling Pt: 248.5 °C (760 mm Hg)
Melting Pt: 89…93 °C
Storage Temperature: Freezer
MDL Number: MFCD00143786
CAS Number: 79069-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR