N-(tert-Butoxycarbonyl)-L-valine N'-methoxy-N'-methylamide 95%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl (1-(methoxy(methyl)amino)-3-methyl-1-oxobutan-2-yl)carbamate, (S)-[1-(Methoxy-methyl-carbamoyl)-2-methyl-propyl]-carbamic acid-tert-butyl ester, N2-{[(1,1-dimethylethyl)oxy]carbonyl}-N1-methyl-N1-(methyloxy)-L-valinamide, tert-butyl (S)-(1-(methoxy(methyl)amino)-3-methyl-1-oxobutan-2-yl)carbamate

A692611-1G A692611-25G A692611-5G
103840-594EA 68 USD
103840-594 76844-814 76844-816 76844-818
N-(tert-Butoxycarbonyl)-L-valine N'-methoxy-N'-methylamide 95%
N-(tert-Butoxycarbonyl)-L-valine N'-methoxy-N'-methylamide

Precaución: for Research Use Only

Formula: C₁₂H₂₄N₂O₄
MW: 260.334 g/mol
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151897
CAS Number: 87694-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR