(S)-tert-Butyl (1-(5-bromopyridin-2-yl)ethyl)carbamate 98%

Supplier: Ambeed
Warning

A323702-100MG A323702-250MG A323702-5G
103825-352EA 284 USD
103825-352 103836-294 77154-568 77154-570 77306-310
(S)-tert-Butyl (1-(5-bromopyridin-2-yl)ethyl)carbamate 98%
(S)-tert-Butyl (1-(5-bromopyridin-2-yl)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₇BrN₂O₂
Boiling Pt: 381.8±32.0 °C (760 mm Hg)
Flash Pt: 184.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD29036411
CAS Number: 915720-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR