(S)-tert-Butyl (1-(4-bromophenyl)ethyl)carbamate 98%
Supplier: Ambeed
|
Warning
|
A181612-10G
A181612-250MG
A181612-1G
A181612-5G
103807-630EA
11
USD
103807-630
103846-006
76974-474
76974-476
77083-536
77083-538
(S)-tert-Butyl (1-(4-bromophenyl)ethyl)carbamate 98%
(S)-tert-Butyl (1-(4-bromophenyl)ethyl)carbamate
Precaución: for Research Use Only
Formula:
C₁₃H₁₈BrNO₂ MW: 300.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11506011 CAS Number: 847728-89-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...