(S)-Pyrrolidin-3-ylmethanol 97%

Supplier: Ambeed
Warning

A249022-25G A249022-1G A249022-5G
103811-850EA 50 USD
103811-850 103851-686 77193-624 77304-470 77304-472
(S)-Pyrrolidin-3-ylmethanol 97%
(S)-Pyrrolidin-3-ylmethanol

Precaución: for Research Use Only

Formula: C₅H₁₁NO
Boiling Pt: 176.1 °C (760 mm Hg)
Flash Pt: 90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09260722
CAS Number: 110013-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR