(S)-Propane-1,2-diamine dihydrochloride 95%

Supplier: Ambeed
Warning

A244302-25G A244302-5G A244302-100G A244302-10G A244302-500G A244302-1G
103811-576EA 5 USD
103811-576 103851-408 77090-612 77090-614 77140-340 77140-342 77193-462 77304-316
(S)-Propane-1,2-diamine dihydrochloride 95%
(S)-Propane-1,2-diamine dihydrochloride

Precaución: for Research Use Only

Formula: C₃H₁₂Cl₂N₂
Melting Pt: 227…229 °C
Storage Temperature: Ambient
MDL Number: MFCD00050706
CAS Number: 19777-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR