(S)-(+)-Nipecotic acid 95%

Supplier: Ambeed
Warning

Synonyms: L-Nipecotic acid, (S)-(+)-3-Piperidinecarboxylic acid, (S)-Piperidine-3-carboxylic acid

A101020-250MG A101020-25G A101020-100G
103795-758EA 10 USD
103795-758 103821-450 77282-642 77282-644 77747-756
(S)-(+)-Nipecotic acid 95%
(S)-(+)-Nipecotic acid

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Boiling Pt: 265.8 °C (760 mm Hg)
Melting Pt: 254 °C
Flash Pt: 114.5 °C
Storage Temperature: Ambient
MDL Number: MFCD01630807
CAS Number: 59045-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR