(S)-Pentan-2-amine hydrochloride 97%

Supplier: Ambeed
Warning

A278710-1G A278710-5G A278710-100MG A278710-10G A278710-250MG
103813-424EA 64 USD
103813-424 103853-368 77093-292 77093-294 77153-786 77153-788 77153-790
(S)-Pentan-2-amine hydrochloride 97%
(S)-Pentan-2-amine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₄ClN
MW: 123.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28166282
CAS Number: 216237-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR