(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulfonate 98%

Supplier: Ambeed
Danger

Synonyms: (S)-(+)-Glycidyl nosylate, (S)-Oxiran-2-ylmethyl 3-nitrobenzenesulphonate, Glycidyl (S)-(+)-3-nitrobenzenesulphonate

A129976-100G A129976-10G A129976-250MG A129976-1G A129976-25G A129976-5G
103801-358EA 9 USD
103801-358 76996-320 76996-322 76996-324 77298-376 77298-378 77298-380
(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulfonate 98%
(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulfonate

Precaución: for Research Use Only

Formula: C₉H₉NO₆S
MW: 259.23 g/mol
Boiling Pt: 432.2 °C (760 mm Hg)
Melting Pt: 61…65 °C
Flash Pt: 215.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064582
CAS Number: 115314-14-2
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR