(S)-N4-(3-Chloro-4-fluorophenyl)-7-((tetrahydrofuran-3-yl)oxy)quinazoline-4,6-diamine 98%

Supplier: Ambeed
Warning

A218083-10G A218083-250MG A218083-25G A218083-1G A218083-5G
103818-688EA 15 USD
103818-688 103849-420 76986-940 76986-942 76986-944 77303-476 77303-478
(S)-N4-(3-Chloro-4-fluorophenyl)-7-((tetrahydrofuran-3-yl)oxy)quinazoline-4,6-diamine 98%
(S)-N4-(3-Chloro-4-fluorophenyl)-7-((tetrahydrofuran-3-yl)oxy)quinazoline-4,6-diamine

Precaución: for Research Use Only

Formula: C₁₈H₁₆ClFN₄O₂
MW: 374.79 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18642925
CAS Number: 314771-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR