(S)-(-)-N,α-Dimethylbenzylamine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-N,α-Dimethylbenzylamine, (S)-(-)-alpha-Methyl-N-methylbenzylamine, (S)-(-)-N-Methyl-1-phenylethylamine, (S)-(-)-N α-Dimethylbenzylamine

A574108-1G A574108-5G A574108-10G
103828-820EA 19 USD
103828-820 103839-962 76844-092 76844-094 77254-092
(S)-(-)-N,α-Dimethylbenzylamine 97%
(S)-(-)-N,α-Dimethylbenzylamine

Precaución: for Research Use Only

Formula: C₉H₁₃N
MW: 135.2100 g/mol
Boiling Pt: 178.7 °C (760 mm Hg)
Flash Pt: 61.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00067113
CAS Number: 19131-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR