(S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine 98%

Supplier: Ambeed
Warning

Synonyms: (S)-N,N-Dimethyl-1-ferrocenylethylamine

A642437-1G A642437-5G A642437-100MG A642437-250MG A642437-25G
103840-350EA 24 USD
103840-350 76844-526 76844-528 77145-266 77145-268 77145-270
(S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine 98%
(S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine

Precaución: for Research Use Only

Formula: C₁₄H₁₉FeN
MW: 257.16 g/mol
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00066272
CAS Number: 31886-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR