(S)-N-(Pyrrolidin-3-yl)acetamide hydrochloride 97%

Supplier: Ambeed
Warning

A185753-1G A185753-5G A185753-250MG
103815-878EA 40 USD
103815-878 103846-448 77084-082 77084-084 77671-622
(S)-N-(Pyrrolidin-3-yl)acetamide hydrochloride 97%
(S)-N-(Pyrrolidin-3-yl)acetamide hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃ClN₂O
MW: 164.63 g/mol
Melting Pt: 127…131 °C
Storage Temperature: Ambient
MDL Number: MFCD16619500
CAS Number: 1246277-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR