(S)-(+)-Methyl-3-hydroxybutyrate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl (S)-(+)-3-Hydroxybutyrate, (S)-Methyl 3-hydroxybutanoate, Methyl (S)-(+)-3-hydroxybutyrate
A488650-100G
A488650-10G
A488650-25G
A488650-1G
A488650-5G
A488650-500G
103828-198EA
5
USD
103828-198
103839-270
76804-858
76804-860
76804-862
76843-222
76843-224
77669-192
(S)-(+)-Methyl-3-hydroxybutyrate 97%
(S)-(+)-Methyl-3-hydroxybutyrate
Precaución: for Research Use Only
Formula:
C₅H₁₀O₃ Boiling Pt: 161.9 °C (760 mm Hg) Flash Pt: 73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064461 CAS Number: 53562-86-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...