N-(Triphenylmethyl)-L-serine methyl ester 95%

Supplier: Ambeed
Warning

Synonyms: (S)-Methyl 3-hydroxy-2-(tritylamino)propanoate, Trt-Ser-OMe, N-Trityl-L-serine Methyl Ester

A412821-100G A412821-10G A412821-250MG A412821-25G A412821-5G A412821-1G
103827-254EA 6 USD
103827-254 103838-300 76725-188 76725-190 76725-192 76841-850 76841-852 77307-160
N-(Triphenylmethyl)-L-serine methyl ester 95%
N-(Triphenylmethyl)-L-serine methyl ester

Precaución: for Research Use Only

Formula: C₂₃H₂₃NO₃
Boiling Pt: 538.2 °C (760 mm Hg)
Melting Pt: 148…150 °C
Flash Pt: 279.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00192570
CAS Number: 4465-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR