(S)-β3-phenylalanine methyl ester hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (S)-methyl 3-amino-3-phenylpropanoate hydrochloride

A110879-100G A110879-10G A110879-250MG A110879-25G A110879-1G A110879-5G
103797-744EA 15 USD
103797-744 103823-552 76821-810 76821-812 76821-814 76821-816 77310-346 77310-348
(S)-β3-phenylalanine methyl ester hydrochloride 97%
(S)-β3-phenylalanine methyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂·ClH
Storage Temperature: Refrigerator
MDL Number: MFCD11052922
CAS Number: 144494-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR