(S)-Methyl 3-amino-2-((tert-butoxycarbonyl)amino)propanoate hydrochloride 95%

Supplier: Ambeed
Warning

A433515-100MG A433515-250MG A433515-1G A433515-5G
103827-554EA 75 USD
103827-554 103838-622 76829-564 76829-566 76842-324 76842-326
(S)-Methyl 3-amino-2-((tert-butoxycarbonyl)amino)propanoate hydrochloride 95%
(S)-Methyl 3-amino-2-((tert-butoxycarbonyl)amino)propanoate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₉ClN₂O₄
MW: 254.71 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04973085
CAS Number: 181228-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR