(S)-Methyl 2-amino-3-cyclohexylpropanoate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A194673-1G
A194673-5G
A194673-100G
A194673-10G
A194673-25G
103816-682EA
26
USD
103816-682
103847-294
77085-216
77085-218
77164-378
77164-380
77164-382
(S)-Methyl 2-amino-3-cyclohexylpropanoate hydrochloride 97%
(S)-Methyl 2-amino-3-cyclohexylpropanoate hydrochloride
Precaución: for Research Use Only
Formula:
C₁₀H₂₀ClNO₂ Melting Pt: 150…154 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00153486 CAS Number: 17193-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...