(S)-Methyl 2-amino-3-(3-hydroxyphenyl)propanoate hydrochloride 97%

Supplier: Ambeed
Warning

A140886-100MG A140886-250MG A140886-5G
103802-914EA 107 USD
103802-914 76984-168 76984-170 77316-032
(S)-Methyl 2-amino-3-(3-hydroxyphenyl)propanoate hydrochloride 97%
(S)-Methyl 2-amino-3-(3-hydroxyphenyl)propanoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClNO₃
MW: 231.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00797541
CAS Number: 34260-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR