(S)-Amino-cyclopentyl-acetic acid methyl ester hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (S)-Methyl 2-amino-2-cyclopentylacetate hydrochloride

A114616-250MG A114616-25G A114616-1G A114616-5G
103798-432EA 28 USD
103798-432 103824-272 76797-028 76797-030 77311-090 77311-092
(S)-Amino-cyclopentyl-acetic acid methyl ester hydrochloride 97%
(S)-Amino-cyclopentyl-acetic acid methyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂·ClH
MW: 193.671 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09800494
CAS Number: 14328-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR