(S)-Methyl 2-amino-2-(4-hydroxyphenyl)acetate hydrochloride 95%

Supplier: Ambeed
Warning

A194104-100MG A194104-250MG A194104-1G A194104-25G A194104-5G
103816-616EA 13 USD
103816-616 103847-226 76974-966 76974-968 77302-388 77302-390 77302-392
(S)-Methyl 2-amino-2-(4-hydroxyphenyl)acetate hydrochloride 95%
(S)-Methyl 2-amino-2-(4-hydroxyphenyl)acetate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO₃
MW: 217.64 g/mol
Melting Pt: 179…183 °C
Storage Temperature: Ambient
MDL Number: MFCD01862153
CAS Number: 127369-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR